Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGGCCAGCTCCTCGTAGTTCA[C/T]GTCAGGACTGGGGACAGGACAGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186852 MIM: 608735 | ||||||||||||||||||||
Literature Links: |
PSMC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMC3 - proteasome 26S subunit, ATPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002804.4 | 1329 | Missense Mutation | ATG,GTG | M,V 379 | NP_002795.2 | |
XM_017018026.1 | 1329 | Missense Mutation | ATG,GTG | M,V 379 | XP_016873515.1 |
SLC39A13 - solute carrier family 39 member 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |