Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGGGAATCAGCACCTCGCGCTC[C/T]TTGGGAAAGACAGAGAAGGCCTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601625 MIM: 610625 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ART1 - ADP-ribosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ART5 - ADP-ribosyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079536.1 | 295 | Silent Mutation | AAA,AAG | K,K 218 | NP_001073004.1 | |
NM_001297668.1 | 295 | Intron | NP_001284597.1 | |||
NM_053017.4 | 295 | Silent Mutation | AAA,AAG | K,K 218 | NP_443750.2 | |
XM_005252779.3 | 295 | Silent Mutation | AAA,AAG | K,K 218 | XP_005252836.1 | |
XM_006718136.3 | 295 | Silent Mutation | AAA,AAG | K,K 218 | XP_006718199.1 | |
XM_011519879.2 | 295 | Silent Mutation | AAA,AAG | K,K 218 | XP_011518181.1 | |
XM_017017171.1 | 295 | Silent Mutation | AAA,AAG | K,K 26 | XP_016872660.1 |
TRPC2 - transient receptor potential cation channel subfamily C member 2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |