Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCGGCGGCGCCGAACAATCCC[C/T]GGCAGGGCTCGCCTCGCCGGTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LUZP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LUZP2 - leucine zipper protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009909.3 | 56 | UTR 5 | NP_001009909.2 | |||
NM_001252008.1 | 56 | UTR 5 | NP_001238937.1 | |||
NM_001252010.1 | 56 | UTR 5 | NP_001238939.1 | |||
XM_011520056.2 | 56 | Missense Mutation | CCG,CTG | P,L 55 | XP_011518358.1 | |
XM_017017648.1 | 56 | Intron | XP_016873137.1 | |||
XM_017017649.1 | 56 | Intron | XP_016873138.1 | |||
XM_017017650.1 | 56 | UTR 5 | XP_016873139.1 | |||
XM_017017651.1 | 56 | UTR 5 | XP_016873140.1 |