Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTAAAATTAAATCTTTGGGGAA[C/T]TATTTTGACATAATTAAATGAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM76B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM76B - family with sequence similarity 76 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144664.4 | 2036 | UTR 3 | NP_653265.3 | |||
XM_005273775.4 | 2036 | UTR 3 | XP_005273832.1 | |||
XM_005273776.4 | 2036 | UTR 3 | XP_005273833.1 | |||
XM_011542610.2 | 2036 | UTR 3 | XP_011540912.1 | |||
XM_011542611.2 | 2036 | UTR 3 | XP_011540913.1 | |||
XM_011542612.1 | 2036 | UTR 3 | XP_011540914.1 | |||
XM_011542613.1 | 2036 | UTR 3 | XP_011540915.1 |