Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCACCAGGCATCTGGAGGCTTC[A/T]TCAATGTTTATATTTTCCTATGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606281 | ||||||||||||||||||||
Literature Links: |
RAB38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB38 - RAB38, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022337.2 | 584 | Missense Mutation | GAA,GAT | E,D 167 | NP_071732.1 | |
XM_017017455.1 | 584 | Intron | XP_016872944.1 | |||
XM_017017456.1 | 584 | Intron | XP_016872945.1 |