Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAAAGTGTAGCAGCACCTTTA[A/C]GCTTAAAAGTGAAGTCAACAAGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615584 | ||||||||||||||||||||
Literature Links: |
FAM111B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM111B - family with sequence similarity 111 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142703.1 | 407 | Missense Mutation | AAG,ACG | K,T 25 | NP_001136175.1 | |
NM_001142704.1 | 407 | Missense Mutation | AAG,ACG | K,T 25 | NP_001136176.1 | |
NM_198947.3 | 407 | Missense Mutation | AAG,ACG | K,T 55 | NP_945185.1 |
LOC101927204 - uncharacterized LOC101927204 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |