Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGAAGCCAAGTCTGCTGTGCC[A/G]GCCCCTGTCCTGCTTCCTTATGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611211 | ||||||||||||||||||||
Literature Links: |
RELT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RELT - RELT tumor necrosis factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032871.3 | 188 | Missense Mutation | CAG,CGG | Q,R 8 | NP_116260.2 | |
NM_152222.1 | 188 | Missense Mutation | CAG,CGG | Q,R 8 | NP_689408.1 | |
XM_005274356.2 | 188 | Intron | XP_005274413.1 | |||
XM_006718714.2 | 188 | UTR 5 | XP_006718777.1 | |||
XM_011545305.1 | 188 | UTR 5 | XP_011543607.1 |