Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTAGTGAGAGCATTGGTACCAC[C/T]TGGTGTGTTCTGAGCTGCACTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601074 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CELF1 - CUGBP, Elav-like family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025596.2 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | NP_001020767.1 | |
NM_001172639.1 | 1094 | Missense Mutation | AGT,GGT | S,G 305 | NP_001166110.1 | |
NM_001172640.1 | 1094 | Missense Mutation | AGT,GGT | S,G 278 | NP_001166111.1 | |
NM_006560.3 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | NP_006551.1 | |
NM_198700.2 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | NP_941989.1 | |
XM_011519848.2 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518150.1 | |
XM_011519850.2 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_011518152.2 | |
XM_011519852.2 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_011518154.1 | |
XM_011519854.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518156.1 | |
XM_011519855.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518157.1 | |
XM_011519856.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518158.1 | |
XM_011519857.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518159.1 | |
XM_011519858.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_011518160.1 | |
XM_011519859.1 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | XP_011518161.1 | |
XM_017017101.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872590.1 | |
XM_017017102.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872591.1 | |
XM_017017103.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872592.1 | |
XM_017017104.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872593.1 | |
XM_017017105.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872594.1 | |
XM_017017106.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872595.1 | |
XM_017017107.1 | 1094 | Missense Mutation | AGT,GGT | S,G 306 | XP_016872596.1 | |
XM_017017108.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872597.1 | |
XM_017017109.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872598.1 | |
XM_017017110.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872599.1 | |
XM_017017111.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872600.1 | |
XM_017017112.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872601.1 | |
XM_017017113.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872602.1 | |
XM_017017114.1 | 1094 | Missense Mutation | AGT,GGT | S,G 302 | XP_016872603.1 | |
XM_017017115.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872604.1 | |
XM_017017116.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872605.1 | |
XM_017017117.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872606.1 | |
XM_017017118.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872607.1 | |
XM_017017119.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872608.1 | |
XM_017017120.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872609.1 | |
XM_017017121.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872610.1 | |
XM_017017122.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872611.1 | |
XM_017017123.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872612.1 | |
XM_017017124.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872613.1 | |
XM_017017125.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872614.1 | |
XM_017017126.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872615.1 | |
XM_017017127.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872616.1 | |
XM_017017128.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872617.1 | |
XM_017017129.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872618.1 | |
XM_017017130.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872619.1 | |
XM_017017131.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872620.1 | |
XM_017017132.1 | 1094 | Missense Mutation | AGT,GGT | S,G 279 | XP_016872621.1 | |
XM_017017133.1 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | XP_016872622.1 | |
XM_017017134.1 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | XP_016872623.1 | |
XM_017017135.1 | 1094 | Missense Mutation | AGT,GGT | S,G 275 | XP_016872624.1 |