Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTTACCTGATTATGAAAGTAT[C/T]AACCATGTTGTGGTTTTTATGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614908 | ||||||||||||||||||||
Literature Links: |
HIKESHI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIKESHI - Hikeshi, heat shock protein nuclear import factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322404.1 | 114 | Silent Mutation | ATC,ATT | I,I 33 | NP_001309333.1 | |
NM_001322407.1 | 114 | UTR 5 | NP_001309336.1 | |||
NM_001322409.1 | 114 | UTR 5 | NP_001309338.1 | |||
NM_016401.3 | 114 | Silent Mutation | ATC,ATT | I,I 33 | NP_057485.2 | |
XM_017017914.1 | 114 | Silent Mutation | ATC,ATT | I,I 33 | XP_016873403.1 | |
XM_017017915.1 | 114 | UTR 5 | XP_016873404.1 |