Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCCGCACCACCACCACCACCA[C/T]CACCTCAGCCCCCACCCGCCGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165360 | ||||||||||||||||||||
Literature Links: |
CBL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBL - Cbl proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005188.3 | 265 | Silent Mutation | CAC,CAT | H,H 41 | NP_005179.2 | |
XM_011543057.2 | 265 | Silent Mutation | CAC,CAT | H,H 41 | XP_011541359.1 | |
XM_017018511.1 | 265 | Intron | XP_016874000.1 |
CCDC153 - coiled-coil domain containing 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145018.1 | 265 | Intron | NP_001138490.1 | |||
XM_006718818.3 | 265 | Intron | XP_006718881.1 | |||
XM_011542763.2 | 265 | Intron | XP_011541065.1 | |||
XM_011542764.2 | 265 | Intron | XP_011541066.1 | |||
XM_011542770.2 | 265 | Intron | XP_011541072.1 | |||
XM_011542771.2 | 265 | Intron | XP_011541073.1 | |||
XM_011542772.2 | 265 | Intron | XP_011541074.1 | |||
XM_011542774.2 | 265 | Intron | XP_011541076.1 | |||
XM_017017577.1 | 265 | Intron | XP_016873066.1 | |||
XM_017017578.1 | 265 | Intron | XP_016873067.1 | |||
XM_017017579.1 | 265 | Intron | XP_016873068.1 | |||
XM_017017580.1 | 265 | Intron | XP_016873069.1 |