Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTCTTCCAGCATCATCAGCATC[A/G]TGCTATTACAATCCCAAACCATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606502 | ||||||||||||||||||||
Literature Links: |
MS4A7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A7 - membrane spanning 4-domains A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021201.4 | 194 | Missense Mutation | ATG,GTG | M,V 1 | NP_067024.1 | |
NM_206938.1 | 194 | Missense Mutation | ATG,GTG | M,V 1 | NP_996821.1 | |
NM_206939.1 | 194 | Missense Mutation | ATG,GTG | M,V 1 | NP_996822.1 | |
NM_206940.1 | 194 | Missense Mutation | ATG,GTG | M,V 1 | NP_996823.1 |