Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTAGACTCTATACCTCCTGCCA[C/T]ACTACCCGTCCCTGCTAGGCAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605473 | ||||||||||||||||||||
Literature Links: |
OR51B5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR51B5 - olfactory receptor family 51 subfamily B member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLN3 - ubiquilin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017481.2 | 1850 | Missense Mutation | ATG,GTG | M,V 555 | NP_059509.1 | |
XM_011520144.2 | 1850 | Missense Mutation | ATG,GTG | M,V 555 | XP_011518446.1 | |
XM_011520145.2 | 1850 | Missense Mutation | ATG,GTG | M,V 402 | XP_011518447.1 |
UBQLNL - ubiquilin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |