Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCTTAAGCACCACCCGTTGAAG[G/T]CAAATGAGCCGTCTTCAGCAGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB13 - DnaJ heat shock protein family (Hsp40) member B13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153614.3 | 1114 | Missense Mutation | GCA,TCA | A,S 36 | NP_705842.2 | |
XM_005273984.3 | 1114 | Missense Mutation | GCA,TCA | A,S 36 | XP_005274041.1 | |
XM_011545004.2 | 1114 | Missense Mutation | GCA,TCA | A,S 70 | XP_011543306.1 | |
XM_011545005.2 | 1114 | Missense Mutation | GCA,TCA | A,S 70 | XP_011543307.1 | |
XM_011545007.2 | 1114 | UTR 5 | XP_011543309.1 | |||
XM_011545009.2 | 1114 | UTR 5 | XP_011543311.1 | |||
XM_011545013.2 | 1114 | Missense Mutation | GCA,TCA | A,S 70 | XP_011543315.1 | |
XM_011545014.2 | 1114 | Missense Mutation | GCA,TCA | A,S 70 | XP_011543316.1 | |
XM_011545015.2 | 1114 | Intron | XP_011543317.1 | |||
XM_017017675.1 | 1114 | UTR 5 | XP_016873164.1 | |||
XM_017017676.1 | 1114 | UTR 5 | XP_016873165.1 | |||
XM_017017677.1 | 1114 | UTR 5 | XP_016873166.1 | |||
XM_017017678.1 | 1114 | UTR 5 | XP_016873167.1 | |||
XM_017017679.1 | 1114 | Missense Mutation | GCA,TCA | A,S 70 | XP_016873168.1 |