Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCCCTCACCGCCCCATTCCATC[C/T]TGTTGTCTTTCCTGGAATTTTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM4B - RNA binding motif protein 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTBN2 - spectrin beta, non-erythrocytic 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006946.2 | 10396 | Intron | NP_008877.1 | |||
XM_005274192.4 | 10396 | UTR 3 | XP_005274249.1 | |||
XM_005274193.3 | 10396 | UTR 3 | XP_005274250.1 | |||
XM_006718669.3 | 10396 | UTR 3 | XP_006718732.1 | |||
XM_006718671.3 | 10396 | UTR 3 | XP_006718734.1 | |||
XM_011545216.2 | 10396 | Intron | XP_011543518.1 | |||
XM_011545217.2 | 10396 | Intron | XP_011543519.1 | |||
XM_017018174.1 | 10396 | UTR 3 | XP_016873663.1 | |||
XM_017018175.1 | 10396 | UTR 3 | XP_016873664.1 | |||
XM_017018176.1 | 10396 | UTR 3 | XP_016873665.1 | |||
XM_017018177.1 | 10396 | UTR 3 | XP_016873666.1 | |||
XM_017018178.1 | 10396 | UTR 3 | XP_016873667.1 |