Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGCTGCTCAAAATTTCTCTTC[C/T]AGGTGCTTAAATCTCCTATCTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606941 | ||||||||||||||||||||
Literature Links: |
ALG9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALG9 - ALG9, alpha-1,2-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C11orf1 - chromosome 11 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022761.2 | 85 | Nonsense Mutation | CAG,TAG | Q,* 15 | NP_073598.1 | |
XM_011542957.2 | 85 | UTR 5 | XP_011541259.1 | |||
XM_011542958.2 | 85 | UTR 5 | XP_011541260.1 | |||
XM_017018129.1 | 85 | Nonsense Mutation | CAG,TAG | Q,* 15 | XP_016873618.1 |
FDXACB1 - ferredoxin-fold anticodon binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138378.2 | 85 | Intron | NP_612387.1 |