Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACTTCCCAGTTCCGCAAGAAC[A/C]GTGGGCGACAGTTATGGAGAAGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPUL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPUL2 - heterogeneous nuclear ribonucleoprotein U like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPUL2-BSCL2 - HNRNPUL2-BSCL2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC9C - tetratricopeptide repeat domain 9C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318812.1 | 270 | UTR 5 | NP_001305741.1 | |||
NM_001318813.1 | 270 | UTR 5 | NP_001305742.1 | |||
NM_001318814.1 | 270 | UTR 5 | NP_001305743.1 | |||
NM_001318815.1 | 270 | UTR 5 | NP_001305744.1 | |||
NM_001318816.1 | 270 | UTR 5 | NP_001305745.1 | |||
NM_173810.3 | 270 | UTR 5 | NP_776171.1 |