Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCGGGAGGTCTGTCAGCCCCA[A/G]CTGTAGAAGTAGATTTTCTGCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604641 MIM: 603360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf94 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf94 - chromosome 11 open reading frame 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP1 - mitogen-activated protein kinase 8 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX16 - peroxisomal biogenesis factor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004813.2 | 1314 | Silent Mutation | AGC,AGT | S,S 334 | NP_004804.1 | |
NM_057174.2 | 1314 | Intron | NP_476515.1 | |||
XM_011520474.1 | 1314 | Silent Mutation | AGC,AGT | S,S 293 | XP_011518776.1 |