Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATCTCAGGATCATCTTCAGGC[A/G]CATACACATTCTGCAAGGTCAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BUD13 - BUD13 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZPR1 - ZPR1 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317086.1 | 1304 | Missense Mutation | GCG,GTG | A,V 365 | NP_001304015.1 | |
NM_003904.4 | 1304 | Missense Mutation | GCG,GTG | A,V 419 | NP_003895.1 |