Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGTCCCAGAAGGAATTGCTCTC[A/G]CTCTGCGATGTCGGGCAATGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602090 MIM: 607982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTBP3 - latent transforming growth factor beta binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130144.2 | 4024 | Silent Mutation | AGC,AGT | S,S 1191 | NP_001123616.1 | |
NM_001164266.1 | 4024 | Silent Mutation | AGC,AGT | S,S 1027 | NP_001157738.1 | |
NM_021070.4 | 4024 | Silent Mutation | AGC,AGT | S,S 1144 | NP_066548.2 | |
XM_011545032.2 | 4024 | Silent Mutation | AGC,AGT | S,S 1200 | XP_011543334.1 | |
XM_011545033.2 | 4024 | Silent Mutation | AGC,AGT | S,S 1153 | XP_011543335.1 | |
XM_017017737.1 | 4024 | Intron | XP_016873226.1 |
SCYL1 - SCY1 like pseudokinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048218.1 | 4024 | Intron | NP_001041683.1 | |||
NM_020680.3 | 4024 | Intron | NP_065731.3 | |||
XM_005274118.3 | 4024 | Intron | XP_005274175.1 | |||
XM_005274120.3 | 4024 | Intron | XP_005274177.1 | |||
XM_005274121.3 | 4024 | Intron | XP_005274178.1 | |||
XM_017018029.1 | 4024 | Intron | XP_016873518.1 | |||
XM_017018030.1 | 4024 | Intron | XP_016873519.1 | |||
XM_017018031.1 | 4024 | Intron | XP_016873520.1 | |||
XM_017018032.1 | 4024 | Intron | XP_016873521.1 | |||
XM_017018033.1 | 4024 | Intron | XP_016873522.1 |