Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCACCTACTCATGCACAAAACT[A/G]CCTCCCAAAGACTTTTCCCAGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIRC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BIRC2 - baculoviral IAP repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166.4 | 1465 | Missense Mutation | ACC,GCC | T,A 5 | NP_001157.1 | |
NM_001256163.1 | 1465 | Missense Mutation | ACC,GCC | T,A 5 | NP_001243092.1 | |
NM_001256166.1 | 1465 | Intron | NP_001243095.1 |