Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTACCATCTTTCATGAAAACT[A/G]CATCAACTTTGTGGCCAATTCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120353 | ||||||||||||||||||||
Literature Links: |
MMP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP1 - matrix metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145938.1 | 1426 | Missense Mutation | GCA,GTA | A,V 362 | NP_001139410.1 | |
NM_002421.3 | 1426 | Missense Mutation | GCA,GTA | A,V 428 | NP_002412.1 |
WTAPP1 - Wilms tumor 1 associated protein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |