Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGGCCTGTTTTCCAAAGTCAC[A/G]CTCATTGGCCGGAGGAAGCTCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605628 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTATIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTATIP2 - HIV-1 Tat interactive protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098520.1 | 210 | Silent Mutation | ACA,ACG | T,T 82 | NP_001091990.1 | |
NM_001098521.1 | 210 | Silent Mutation | ACA,ACG | T,T 48 | NP_001091991.1 | |
NM_001098522.1 | 210 | Silent Mutation | ACA,ACG | T,T 48 | NP_001091992.1 | |
NM_001098523.1 | 210 | Silent Mutation | ACA,ACG | T,T 48 | NP_001091993.1 | |
NM_006410.4 | 210 | Silent Mutation | ACA,ACG | T,T 48 | NP_006401.3 |