Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGGTGAATGAGGCCTCTGGGG[A/G]TGGGGATGGAGAGGATGCCGTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCPS - decapping enzyme, scavenger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014026.4 | 1001 | Missense Mutation | GAT,GGT | D,G 74 | NP_054745.1 | |
XM_011542778.2 | 1001 | Missense Mutation | GAT,GGT | D,G 81 | XP_011541080.1 | |
XM_011542780.2 | 1001 | Intron | XP_011541082.1 | |||
XM_017017612.1 | 1001 | UTR 5 | XP_016873101.1 |
HP11014 - uncharacterized HP11014 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |