Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAAAGTTTCCAGCCCTTGGTAA[C/T]GCCCAGACAAAGGTCCTTCCCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGAP2 - ADP ribosylation factor GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242832.1 | 2852 | UTR 3 | NP_001229761.1 | |||
NM_032389.4 | 2852 | UTR 3 | NP_115765.2 | |||
XM_005253166.1 | 2852 | UTR 3 | XP_005253223.1 | |||
XM_005253167.1 | 2852 | UTR 3 | XP_005253224.1 | |||
XM_005253168.2 | 2852 | UTR 3 | XP_005253225.1 | |||
XM_006718346.1 | 2852 | UTR 3 | XP_006718409.1 | |||
XM_011520405.1 | 2852 | UTR 3 | XP_011518707.1 | |||
XM_017018413.1 | 2852 | UTR 3 | XP_016873902.1 | |||
XM_017018414.1 | 2852 | UTR 3 | XP_016873903.1 | |||
XM_017018415.1 | 2852 | UTR 3 | XP_016873904.1 |
C11orf49 - chromosome 11 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003676.2 | 2852 | Intron | NP_001003676.1 | |||
NM_001003677.2 | 2852 | Intron | NP_001003677.1 | |||
NM_001003678.2 | 2852 | Intron | NP_001003678.1 | |||
NM_001278222.1 | 2852 | Intron | NP_001265151.1 | |||
NM_024113.4 | 2852 | Intron | NP_077018.1 | |||
XM_006718315.1 | 2852 | Intron | XP_006718378.1 | |||
XM_011520364.2 | 2852 | Intron | XP_011518666.1 | |||
XM_011520365.2 | 2852 | Intron | XP_011518667.1 | |||
XM_017018276.1 | 2852 | Intron | XP_016873765.1 | |||
XM_017018277.1 | 2852 | Intron | XP_016873766.1 | |||
XM_017018278.1 | 2852 | Intron | XP_016873767.1 | |||
XM_017018279.1 | 2852 | Intron | XP_016873768.1 | |||
XM_017018280.1 | 2852 | Intron | XP_016873769.1 | |||
XM_017018281.1 | 2852 | Intron | XP_016873770.1 | |||
XM_017018282.1 | 2852 | Intron | XP_016873771.1 |