Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAATAGACAGGAAACCCAAGTT[A/T]GCCCAATTTCTGGGCGTGGGAATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607424 | ||||||||||||||||||||
Literature Links: |
GLYAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLYAT - glycine-N-acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005838.3 | 984 | Intron | NP_005829.3 | |||
NM_201648.2 | 984 | Missense Mutation | AAA,TAA | K,* 256 | NP_964011.2 | |
XM_017017087.1 | 984 | Nonsense Mutation | AAA,TAA | K,* 192 | XP_016872576.1 |