Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTCGAAGTCTGTCTTTCTCTAC[C/T]TGTAAGCGTTCCTTTTCAATCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MSANTD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSANTD4 - Myb/SANT DNA binding domain containing 4 with coiled-coils | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318747.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | NP_001305676.1 | |
NM_001318748.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | NP_001305677.1 | |
NM_001318749.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | NP_001305678.1 | |
NM_001318750.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | NP_001305679.1 | |
NM_032424.2 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | NP_115800.1 | |
XM_011543022.2 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | XP_011541324.1 | |
XM_017018416.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | XP_016873905.1 | |
XM_017018417.1 | 1592 | Silent Mutation | CAA,CAG | Q,Q 330 | XP_016873906.1 |