Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGAGTGAGGCTGGAGAGGGGCT[C/T]GGCAATCTTCAGGAATTGATGCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602590 | ||||||||||||||||||||
Literature Links: |
PAK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAK1 - p21 (RAC1) activated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128620.1 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | NP_001122092.1 | |
NM_002576.4 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | NP_002567.3 | |
XM_011545080.2 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | XP_011543382.1 | |
XM_011545081.2 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | XP_011543383.1 | |
XM_011545082.2 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | XP_011543384.1 | |
XM_011545083.2 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | XP_011543385.1 | |
XM_011545084.2 | 1822 | Missense Mutation | CAA,CGA | Q,R 541 | XP_011543386.1 | |
XM_017017846.1 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | XP_016873335.1 | |
XM_017017847.1 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | XP_016873336.1 | |
XM_017017848.1 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | XP_016873337.1 | |
XM_017017849.1 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | XP_016873338.1 | |
XM_017017850.1 | 1822 | Missense Mutation | AAG,GAG | K,E 525 | XP_016873339.1 |