Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGCCCACCCCAATCACAAACTA[G/T]AAGCAGCGTTGTACAAACAGGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608707 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDON PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDON - cell adhesion associated, oncogene regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243597.1 | 7495 | UTR 3 | NP_001230526.1 | |||
NM_016952.4 | 7495 | UTR 3 | NP_058648.4 | |||
XM_011542862.2 | 7495 | UTR 3 | XP_011541164.1 | |||
XM_011542863.2 | 7495 | UTR 3 | XP_011541165.1 | |||
XM_011542864.2 | 7495 | UTR 3 | XP_011541166.1 | |||
XM_011542865.2 | 7495 | UTR 3 | XP_011541167.1 | |||
XM_011542866.2 | 7495 | UTR 3 | XP_011541168.1 | |||
XM_017017873.1 | 7495 | UTR 3 | XP_016873362.1 |
LOC338667 - V-set and immunoglobulin domain-containing protein 10-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |