Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGATGATGGGGGCTGTGACCAC[A/G]CTCTTGCCCTTCATTGGCTGGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164013 | ||||||||||||||||||||
Literature Links: |
NFRKB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFRKB - nuclear factor related to kappaB binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143835.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1199 | NP_001137307.1 | |
NM_006165.3 | 3709 | Silent Mutation | AGC,AGT | S,S 1224 | NP_006156.2 | |
XM_005271567.3 | 3709 | Silent Mutation | AGC,AGT | S,S 1211 | XP_005271624.1 | |
XM_005271568.3 | 3709 | Silent Mutation | AGC,AGT | S,S 1211 | XP_005271625.1 | |
XM_011542850.2 | 3709 | Silent Mutation | AGC,AGT | S,S 1234 | XP_011541152.1 | |
XM_011542851.2 | 3709 | Silent Mutation | AGC,AGT | S,S 1224 | XP_011541153.1 | |
XM_011542852.2 | 3709 | Silent Mutation | AGC,AGT | S,S 1224 | XP_011541154.1 | |
XM_017017793.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1241 | XP_016873282.1 | |
XM_017017794.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1229 | XP_016873283.1 | |
XM_017017795.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1222 | XP_016873284.1 | |
XM_017017796.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1199 | XP_016873285.1 | |
XM_017017797.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1152 | XP_016873286.1 | |
XM_017017798.1 | 3709 | Silent Mutation | AGC,AGT | S,S 1110 | XP_016873287.1 | |
XM_017017799.1 | 3709 | Silent Mutation | AGC,AGT | S,S 956 | XP_016873288.1 | |
XM_017017800.1 | 3709 | Silent Mutation | AGC,AGT | S,S 956 | XP_016873289.1 | |
XM_017017801.1 | 3709 | Silent Mutation | AGC,AGT | S,S 660 | XP_016873290.1 |
TMEM45B - transmembrane protein 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |