Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTAAATTCTAAACACCTTTT[C/T]GTTTGTAGGCTGGGCCAGTAATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf71 - chromosome 11 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM7 - RNA binding motif protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286045.1 | Intron | NP_001272974.1 | ||||
NM_001286046.1 | Intron | NP_001272975.1 | ||||
NM_001286047.1 | Intron | NP_001272976.1 | ||||
NM_001286048.1 | Intron | NP_001272977.1 | ||||
NM_016090.3 | Intron | NP_057174.1 |