Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGTTGCCTTCAGGGTACCTGCT[C/T]TTCCGAGACTTCTGCCTGAACCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109635 | ||||||||||||||||||||
Literature Links: |
GRK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRK2 - G protein-coupled receptor kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001619.3 | 467 | Silent Mutation | CTC,CTT | L,L 67 | NP_001610.2 | |
XM_011544773.1 | 467 | Silent Mutation | CTC,CTT | L,L 37 | XP_011543075.1 |