Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCCCACGAAGGTCGGCTTGCA[C/T]ACATACACCAGGCTGTGGCTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605416 MIM: 614460 | ||||||||||||||||||||
Literature Links: |
DKK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DKK3 - dickkopf WNT signaling pathway inhibitor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018057.1 | 901 | Silent Mutation | GTA,GTG | V,V 283 | NP_001018067.1 | |
NM_013253.4 | 901 | Silent Mutation | GTA,GTG | V,V 283 | NP_037385.2 | |
NM_015881.5 | 901 | Silent Mutation | GTA,GTG | V,V 283 | NP_056965.3 | |
XM_006718178.2 | 901 | Silent Mutation | GTA,GTG | V,V 283 | XP_006718241.1 | |
XM_017017553.1 | 901 | Silent Mutation | GTA,GTG | V,V 297 | XP_016873042.1 | |
XM_017017554.1 | 901 | Silent Mutation | GTA,GTG | V,V 297 | XP_016873043.1 | |
XM_017017555.1 | 901 | Silent Mutation | GTA,GTG | V,V 297 | XP_016873044.1 |
USP47 - ubiquitin specific peptidase 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |