Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGAGAGCAACCGGACAATATA[C/G]CAGCTTTTGCAGCAGCCTATTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610079 MIM: 608621 | ||||||||||||||||||||
Literature Links: |
SIAE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIAE - sialic acid acetylesterase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199922.1 | 259 | UTR 5 | NP_001186851.1 | |||
NM_170601.4 | 259 | Intron | NP_733746.1 | |||
XM_017017930.1 | 259 | Intron | XP_016873419.1 |
SPA17 - sperm autoantigenic protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017425.3 | 259 | Missense Mutation | CCA,GCA | P,A 36 | NP_059121.1 | |
XM_011542870.2 | 259 | Missense Mutation | CCA,GCA | P,A 36 | XP_011541172.1 |