Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGTGTAGAAGATCTCGTCGTAG[A/G]TGGGCTTGTCCTTGCCCACCACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613991 MIM: 605888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42BPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42BPG - CDC42 binding protein kinase gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EHD1 - EH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282444.1 | 1465 | Missense Mutation | ACC,ATC | T,I 447 | NP_001269373.1 | |
NM_001282445.1 | 1465 | Missense Mutation | ACC,ATC | T,I 461 | NP_001269374.1 | |
NM_006795.3 | 1465 | Missense Mutation | ACC,ATC | T,I 447 | NP_006786.2 | |
XM_011544739.1 | 1465 | Missense Mutation | ACC,ATC | T,I 447 | XP_011543041.1 |