Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACTTACATTTTCAATTGCCAGG[A/G]AAGAGGTAGAAATCTCTTGTCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602665 | ||||||||||||||||||||
Literature Links: |
CASP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP5 - caspase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136109.1 | 899 | Silent Mutation | TTC,TTT | F,F 373 | NP_001129581.1 | |
NM_001136110.1 | 899 | Silent Mutation | TTC,TTT | F,F 289 | NP_001129582.1 | |
NM_001136112.1 | 899 | Silent Mutation | TTC,TTT | F,F 444 | NP_001129584.1 | |
NM_004347.3 | 899 | Silent Mutation | TTC,TTT | F,F 431 | NP_004338.3 | |
XM_011543021.2 | 899 | Intron | XP_011541323.1 |