Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180465 MIM: 602671 MIM: 610971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC84 - coiled-coil domain containing 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3656 - microRNA 3656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS25 - ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC37A4 - solute carrier family 37 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC4 - trafficking protein particle complex 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318486.1 | 748 | Intron | NP_001305415.1 | |||
NM_001318488.1 | 748 | Missense Mutation | ACA,GCA | T,A 141 | NP_001305417.1 | |
NM_001318489.1 | 748 | Missense Mutation | ACA,GCA | T,A 87 | NP_001305418.1 | |
NM_001318490.1 | 748 | Missense Mutation | ACA,GCA | T,A 98 | NP_001305419.1 | |
NM_001318492.1 | 748 | Missense Mutation | ACA,GCA | T,A 36 | NP_001305421.1 | |
NM_001318494.1 | 748 | Intron | NP_001305423.1 | |||
NM_016146.5 | 748 | Missense Mutation | ACA,GCA | T,A 141 | NP_057230.1 |