Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGGCGGAGGCTTCGAGTGACC[C/T]GGGTGCCGAGGAGCGGGAAGAGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153432 | ||||||||||||||||||||
Literature Links: |
LSP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LSP1 - lymphocyte-specific protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013253.1 | 198 | Intron | NP_001013271.1 | |||
NM_001013254.1 | 198 | Intron | NP_001013272.1 | |||
NM_001013255.1 | 198 | Intron | NP_001013273.1 | |||
NM_001242932.1 | 198 | Intron | NP_001229861.1 | |||
NM_001289005.1 | 198 | Intron | NP_001275934.1 | |||
NM_002339.2 | 198 | Missense Mutation | CCG,CTG | P,L 8 | NP_002330.1 |
MIR4298 - microRNA 4298 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |