Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTGCAAATACTTGGGGTCCTTC[A/G]TCCATCACTGTTTTGCATAAGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603954 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZW10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZW10 - zw10 kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004724.3 | 1918 | Silent Mutation | GAC,GAT | D,D 691 | NP_004715.1 | |
XM_017018558.1 | 1918 | Silent Mutation | GAC,GAT | D,D 627 | XP_016874047.1 | |
XM_017018559.1 | 1918 | Silent Mutation | GAC,GAT | D,D 471 | XP_016874048.1 |