Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC4 - DnaJ heat shock protein family (Hsp40) member C4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FERMT3 - fermitin family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT22 - nudix hydrolase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128612.2 | 296 | Intron | NP_001122084.1 | |||
NM_001128613.2 | 296 | Intron | NP_001122085.1 | |||
NM_001271831.1 | 296 | Intron | NP_001258760.1 | |||
NM_032344.3 | 296 | Intron | NP_115720.1 |
TRPT1 - tRNA phosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033678.3 | 296 | Missense Mutation | AAC,GAC | N,D 28 | NP_001028850.2 | |
NM_001160389.1 | 296 | Missense Mutation | AAC,GAC | N,D 28 | NP_001153861.1 | |
NM_001160390.1 | 296 | Missense Mutation | AAC,GAC | N,D 28 | NP_001153862.1 | |
NM_001160392.1 | 296 | Missense Mutation | AAC,GAC | N,D 28 | NP_001153864.1 | |
NM_001160393.1 | 296 | Missense Mutation | AAC,GAC | N,D 28 | NP_001153865.1 | |
NM_031472.3 | 296 | UTR 5 | NP_113660.1 | |||
XM_005274345.4 | 296 | Missense Mutation | AAC,GAC | N,D 28 | XP_005274402.1 | |
XM_005274346.4 | 296 | Missense Mutation | AAC,GAC | N,D 28 | XP_005274403.1 | |
XM_005274347.4 | 296 | UTR 5 | XP_005274404.1 | |||
XM_006718706.3 | 296 | Missense Mutation | AAC,GAC | N,D 28 | XP_006718769.1 | |
XM_006718707.3 | 296 | Missense Mutation | AAC,GAC | N,D 28 | XP_006718770.1 | |
XM_017018400.1 | 296 | UTR 5 | XP_016873889.1 | |||
XM_017018401.1 | 296 | UTR 5 | XP_016873890.1 |
VEGFB - vascular endothelial growth factor B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |