Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGTCTCATGCTATCTTGAGCG[G/T]CACTCTGTCACTTCCAAGATGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612793 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTDSS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTDSS2 - phosphatidylserine synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253153.1 | 2443 | Intron | XP_005253210.1 | |||
XM_011520391.2 | 2443 | Intron | XP_011518693.1 | |||
XM_011520392.2 | 2443 | Intron | XP_011518694.1 | |||
XM_011520393.2 | 2443 | Intron | XP_011518695.1 | |||
XM_017018368.1 | 2443 | UTR 5 | XP_016873857.1 | |||
XM_017018369.1 | 2443 | Intron | XP_016873858.1 | |||
XM_017018370.1 | 2443 | Intron | XP_016873859.1 | |||
XM_017018371.1 | 2443 | Intron | XP_016873860.1 |