Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTATGCAAGGGGAGCAGAAGTAT[A/G]TTCCTCCAGAACCGCTCAATAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615680 MIM: 147678 | ||||||||||||||||||||
Literature Links: |
CARD16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARD16 - caspase recruitment domain family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017534.1 | 591 | Silent Mutation | AAC,AAT | N,N 167 | NP_001017534.1 | |
NM_052889.2 | 591 | UTR 3 | NP_443121.1 | |||
XM_011542583.2 | 591 | Silent Mutation | AAC,AAT | N,N 235 | XP_011540885.1 |
CASP1 - caspase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |