Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGGAGGCCCCCCCGTGAGCTT[C/T]GCAGTTGCTTGAGGTCATCCCACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604270 | ||||||||||||||||||||
Literature Links: |
LRP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRP4 - LDL receptor related protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002334.3 | 4802 | Missense Mutation | CAA,CGA | Q,R 1830 | NP_002325.2 | |
XM_011520103.2 | 4802 | Missense Mutation | CAA,CGA | Q,R 1562 | XP_011518405.1 | |
XM_011520104.2 | 4802 | Missense Mutation | CAA,CGA | Q,R 1085 | XP_011518406.1 | |
XM_017017734.1 | 4802 | Intron | XP_016873223.1 |
LRP4-AS1 - LRP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |