Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATGCTATTTGGTGGGAACAGA[A/C]GAGGCAGTGGCTGCTGCAGACCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607901 MIM: 605063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FERMT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FERMT3 - fermitin family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031471.5 | 355 | Missense Mutation | AAG,ACG | K,T 69 | NP_113659.3 | |
NM_178443.2 | 355 | Missense Mutation | AAG,ACG | K,T 69 | NP_848537.1 | |
XM_011545294.2 | 355 | Missense Mutation | AAG,ACG | K,T 69 | XP_011543596.1 | |
XM_011545295.2 | 355 | UTR 5 | XP_011543597.1 | |||
XM_017018398.1 | 355 | Missense Mutation | AAG,ACG | K,T 69 | XP_016873887.1 | |
XM_017018399.1 | 355 | UTR 5 | XP_016873888.1 |
STIP1 - stress induced phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |