Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCCTCTGACTTGGCATACAG[C/G]GGCAGGTTGCAGTAGTCACCTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611782 | ||||||||||||||||||||
Literature Links: |
DSCAML1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSCAML1 - DS cell adhesion molecule like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020693.3 | 4165 | Silent Mutation | CCC,CCG | P,P 1962 | NP_065744.2 | |
XM_011542917.1 | 4165 | Silent Mutation | CCC,CCG | P,P 1914 | XP_011541219.1 | |
XM_011542918.1 | 4165 | Silent Mutation | CCC,CCG | P,P 1826 | XP_011541220.1 | |
XM_011542919.2 | 4165 | Silent Mutation | CCC,CCG | P,P 1475 | XP_011541221.1 | |
XM_011542920.2 | 4165 | Silent Mutation | CCC,CCG | P,P 1392 | XP_011541222.1 | |
XM_011542921.2 | 4165 | Silent Mutation | CCC,CCG | P,P 1193 | XP_011541223.1 | |
XM_011542924.2 | 4165 | Silent Mutation | CCC,CCG | P,P 1119 | XP_011541226.1 | |
XM_011542925.2 | 4165 | Silent Mutation | CCC,CCG | P,P 1113 | XP_011541227.1 |