Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTCTCAGCATGAACTGCATAT[A/C]CGACTTCTTCACCTATGAGACCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600843 MIM: 604328 | ||||||||||||||||||||
Literature Links: |
P2RX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
P2RX3 - purinergic receptor P2X 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002559.3 | 733 | Missense Mutation | TAC,TCC | Y,S 5 | NP_002550.2 | |
XM_011545069.2 | 733 | Missense Mutation | TAC,TCC | Y,S 5 | XP_011543371.1 | |
XM_011545070.2 | 733 | Missense Mutation | TAC,TCC | Y,S 5 | XP_011543372.1 | |
XM_011545071.2 | 733 | Missense Mutation | TAC,TCC | Y,S 5 | XP_011543373.1 | |
XM_011545072.2 | 733 | UTR 5 | XP_011543374.1 | |||
XM_011545073.1 | 733 | Intron | XP_011543375.1 |
SSRP1 - structure specific recognition protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |