Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGATGACAGCAGAGCCTGCACC[A/G]GCTTTGGGGCCCAGACGTTGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176258 MIM: 606051 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNC1 - potassium voltage-gated channel subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERGEF - secretion regulating guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012139.3 | 1282 | Missense Mutation | CCG,CTG | P,L 377 | NP_036271.1 |