Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGCATAATGGCAGCGTCTGAG[G/T]TTGCTGGTGTTGTGGCCAATGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCSK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCSK7 - proprotein convertase subtilisin/kexin type 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF214 - ring finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077239.1 | 124 | Missense Mutation | GTT,TTT | V,F 6 | NP_001070707.1 | |
NM_001278249.1 | 124 | Missense Mutation | GTT,TTT | V,F 6 | NP_001265178.1 | |
NM_207343.3 | 124 | Missense Mutation | GTT,TTT | V,F 6 | NP_997226.2 |