Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACCAGATGTCAACCTTGGGCCC[A/G]TAGGCCTTTCGTGTCACAACCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAK1 - p21 (RAC1) activated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128620.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | NP_001122092.1 | |
NM_002576.4 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | NP_002567.3 | |
XM_011545080.2 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_011543382.1 | |
XM_011545081.2 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_011543383.1 | |
XM_011545082.2 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_011543384.1 | |
XM_011545083.2 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_011543385.1 | |
XM_011545084.2 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_011543386.1 | |
XM_017017846.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016873335.1 | |
XM_017017847.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016873336.1 | |
XM_017017848.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016873337.1 | |
XM_017017849.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016873338.1 | |
XM_017017850.1 | 1572 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016873339.1 |