Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGACCGAGTAGCGTAGCTTACC[A/G]AAGTAACCGCCTTCACCTGGCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM10 - PR domain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020228.2 | 5270 | Missense Mutation | TCG,TTG | S,L 1093 | NP_064613.2 | |
NM_199437.1 | 5270 | Missense Mutation | TCG,TTG | S,L 1089 | NP_955469.1 | |
NM_199438.1 | 5270 | Missense Mutation | TCG,TTG | S,L 994 | NP_955470.1 | |
NM_199439.1 | 5270 | Missense Mutation | TCG,TTG | S,L 956 | NP_955471.1 | |
XM_011542912.2 | 5270 | Missense Mutation | TCG,TTG | S,L 1076 | XP_011541214.1 | |
XM_017018023.1 | 5270 | Missense Mutation | TCG,TTG | S,L 990 | XP_016873512.1 |