Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGGCAGCCTGTGGGCCAGGGC[A/C]CCAGGGGGCAGGCGGTGGTAGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606259 MIM: 605965 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRMS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRMS1 - breast cancer metastasis suppressor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIN1 - Ras and Rab interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004292.2 | 2537 | Silent Mutation | NP_004283.2 | |||
XM_011545399.2 | 2537 | Silent Mutation | XP_011543701.1 | |||
XM_011545400.2 | 2537 | Silent Mutation | XP_011543702.1 | |||
XM_011545401.2 | 2537 | Silent Mutation | XP_011543703.1 | |||
XM_017018587.1 | 2537 | Silent Mutation | XP_016874076.1 |